Learn More
Agilent Technologies GLYCOSEP R HPLC COLUMN
Catalog No. NC9421472
Encompass
A reverse phase column used for optimal separation of fluorescent Signal labeled carbohydrates. Used for profiling of 2-AB-labeled neutral oligosaccharides (single or 2nd dimension after GlycoSep C), high resolution separation of 2-AA-labeled monosaccharides and identification of DMB-labeled sialic acids.
Catalog No. NC9421472 Supplier Agilent Technologies Supplier No. GKI4727
A reverse phase column used for optimal separation of fluorescent Signal labeled carbohydrates. Used for profiling of 2-AB-labeled neutral oligosaccharides (single or 2nd dimension after GlycoSep C), high resolution separation of 2-AA-labeled monosaccharides and identification of DMB-labeled sialic acids.