Learn More
QA Bio Inc QA-Bio, Inc. • Enzymatic DeGlycoMx Deglycosylation Kit • 20 µls
Catalog No. 50355826
Encompass
DeGlycoMx Glycan Deglycosylation Kit • The DeGlycoMx is a premixed cocktail of enzymes deglycoylate all N-linked oligosaccharides and most O-linked sugars.The kit is easy-to-use• add 2 µl of the DeGlycoMx to your sample and buffer, incubate for 3 hours at 37˚C.
Catalog No. 50-355-826 Supplier QA Bio Inc Supplier No. KEDGMX
DeGlycoMx Glycan Deglycosylation Kit • The DeGlycoMx is a premixed cocktail of enzymes deglycoylate all N-linked oligosaccharides and most O-linked sugars.The kit is easy-to-use• add 2 µl of the DeGlycoMx to your sample and buffer, incubate for 3 hours at 37˚C.