Learn More
QA Bio Inc QA-Bio, Inc. • Endo F Multi-Kit Glycan Deglycosylation Kit • 20 µls each of Endo F1, F2, & F3
Catalog No. 50355827
Encompass
Endo F Glycan Deglycosylation Kit • The Endo F Multi-Kit will deglycosylate N-linked glycans in native and denatured conditions. Use the three enzymes in combination to completely remove all N-linked glycans, or use each enzyme alone to determine the type of N-glycans.
Catalog No. 50-355-827 Supplier QA Bio Inc Supplier No. KEEFX3
Endo F Glycan Deglycosylation Kit • The Endo F Multi-Kit will deglycosylate N-linked glycans in native and denatured conditions. Use the three enzymes in combination to completely remove all N-linked glycans, or use each enzyme alone to determine the type of N-glycans.