Learn More
QA Bio Inc QA-Bio, Inc. • PNGase F • native Elizabethkingia miricola • 200 µls
Catalog No. NC1553890
Encompass
PNGase F - 200 µls -E. miricola • Removes N-linked Glycans • No Glycerol, EDTA, or NaCl - Enzyme remains fully active for 4+ days at 37˚C. • Used by Biopharmaceutical Labs Around the World in FDA/EMEA Assays since 2002.
Catalog No. NC1553890 Supplier QA Bio Inc Supplier No. EPNG01200
PNGase F - 200 µls -E. miricola • Removes N-linked Glycans • No Glycerol, EDTA, or NaCl - Enzyme remains fully active for 4+ days at 37˚C. • Used by Biopharmaceutical Labs Around the World in FDA/EMEA Assays since 2002.