Learn More
QA Bio Inc QA-Bio, Inc. • Neuraminidase Au • recombinant from A. ureafaciens • 60 µls
Catalog No. 50355821
Encompass
Neuraminidase Au - 60 µls • Alpha (2-3,6,8,9) Neuraminidase (Sialidase) cleaves all non-reducing terminal neuraminic acid residues from complex carbohydrates and glycoproteins.
Catalog No. 50-355-821 Supplier QA Bio Inc Supplier No. ES001
Neuraminidase Au - 60 µls • Alpha (2-3,6,8,9) Neuraminidase (Sialidase) cleaves all non-reducing terminal neuraminic acid residues from complex carbohydrates and glycoproteins.