Learn More
Advanced Biomatrix Glycosil Version 2.0 Thiol-Modified Hyaluronan, 5 units x 1 mL
Supplier:  Advanced Biomatrix GS222F5EA
Encompass_Preferred
Glycosil is a thiol-modified hyaluronic acid. Glycosil hyaluronic acid is a component of the HyStem, HyStem-C and HyStem-HP hydrogel kits but can be purchased separately here. Comes with 10 mL of Buffer A. HyStem V2.0 kit components are reconstituted with a buffered 1X PBS. The resulting components will also result in a pH neutral isotonic salt hydrogel. The ending pH, osmo, and rheological properties are the same from Version 1.0 to Version 2.0. Glycosil hyaluronic acid is a component of the HyStem and Hystem-C hydrogel kits and can be purchased separately in individual vials. Glycosil is a thiol-modified hyaluronic acid and a constituent of native extracellular matrix (ECM). Glycosil is used in conjunction with Gelin-S or ECM proteins such as laminin, collagen, or fibronectin for most 3-D cell culture and tissue-engineering applications.
Catalog No. NC1728602
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.