Learn More
Cayman Chemical Glycoursodeoxycholc ACDd4 10mg
SDP
Catalog No. 502990775
Encompass_Preferred
An internal standard for the quantification of glycoursodeoxycholic acid by GC- or LC-MS
Catalog No. 50-299-0775 Supplier Cayman Chemical Supplier No. 3130910 MG
An internal standard for the quantification of glycoursodeoxycholic acid by GC- or LC-MS