Learn More
Cayman Chemical Glycoursodeoxycholic Acid, Purity: ≥98%, CAS: 64480-66-6, 500 mg
SDP
Catalog No. 502986325
Encompass_Preferred
A glycine-conjugated form of ursodeoxycholic acid; has antioxidant effects in vitro in Barrett’s esophagus cells and primary cultured rat neurons; reduces the levels of inflammatory cytokines and prevents cell death induced by unconjugated bilirubin in an astroglial cell model of neonatal hyperbilirubinemia; decreases the severity of symptoms and increases the amount of A. muciniphila in a mouse model of colitis at 500 mg/kg per day
Catalog No. 50-298-6325 Supplier Cayman Chemical Supplier No. 21698500 MG
A glycine-conjugated form of ursodeoxycholic acid; has antioxidant effects in vitro in Barrett’s esophagus cells and primary cultured rat neurons; reduces the levels of inflammatory cytokines and prevents cell death induced by unconjugated bilirubin in an astroglial cell model of neonatal hyperbilirubinemia; decreases the severity of symptoms and increases the amount of A. muciniphila in a mouse model of colitis at 500 mg/kg per day