Learn More
Cayman Chemical GlycoursodeoxycholIc ACD Max
SDP
Catalog No. 502978696
Encompass_Preferred
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications
Catalog No. 50-297-8696 Supplier Cayman Chemical Supplier No. 31602100 UG
A quantitative analytical standard guaranteed to meet MaxSpec identity, purity, stability, and concentration specifications