Learn More
Waters Corp Waters N-Glycan Kit, RapiFluor-MS, 96 Sample
Catalog No. 501384487 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies complicated and time-consuming sample preparation into three easy steps that can be completed in as little as 30 minutes. The RapiFluor-MS label offers increased fluorescence quantification and superior mass spectral response, allowing for a single label to provide valuable information from characterization to routine monitoring. This core kit contains materials to process 96 samples in a 4x24 format.
  • Streamlined, fast, and highly sensitive analysis of released N-glycans.
  • Sample preparation in as little as 30 minutes through 3 easy steps.
  • Label provides increased fluorescence quantification and superior mass spectral response.
  • Utilizes a single label for characterization and routine monitoring.
  • Processes 96 samples in a 4x24 format.
Catalog No. 50-138-4487 Supplier Waters Corp Supplier No. 176003606
Add to Cart
Add to Cart
Request a Quote
This kit provides a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies complicated and time-consuming sample preparation into three easy steps that can be completed in as little as 30 minutes. The RapiFluor-MS label offers increased fluorescence quantification and superior mass spectral response, allowing for a single label to provide valuable information from characterization to routine monitoring. This core kit contains materials to process 96 samples in a 4x24 format.
  • Streamlined, fast, and highly sensitive analysis of released N-glycans.
  • Sample preparation in as little as 30 minutes through 3 easy steps.
  • Label provides increased fluorescence quantification and superior mass spectral response.
  • Utilizes a single label for characterization and routine monitoring.
  • Processes 96 samples in a 4x24 format.