Learn More
Waters Corp Waters Corporation GlycoWorks RapiFluor-MS N-Glycan Starter Kit, 96 Sample, HILIC, MS/MS, UV, HRMS Detection
Catalog No. 501384488 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This starter kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies sample preparation to three easy steps, achievable in as little as 30 minutes. The RapiFluor-MS reagent provides increased fluorescence quantification and mass spectral response, enabling valuable information from characterization to routine monitoring for 96 samples.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Three easy sample preparation steps in as little as 30 minutes.
  • RapiFluor-MS reagent for increased fluorescence quantification and mass spectral response.
  • Processes 96 samples in a 4x24 format.
  • Includes modules for deglycosylation, labeling, clean-up, and sample collection.
  • Optimized for glycoprotein quantity up to 40 μg.
  • Rapid workflow: deglycosylation <15 min, labeling 5 min, clean-up 10 min.
Catalog No. 50-138-4488 Supplier Waters Corp Supplier No. 176003635
Add to Cart
Add to Cart
Request a Quote
This starter kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies sample preparation to three easy steps, achievable in as little as 30 minutes. The RapiFluor-MS reagent provides increased fluorescence quantification and mass spectral response, enabling valuable information from characterization to routine monitoring for 96 samples.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Three easy sample preparation steps in as little as 30 minutes.
  • RapiFluor-MS reagent for increased fluorescence quantification and mass spectral response.
  • Processes 96 samples in a 4x24 format.
  • Includes modules for deglycosylation, labeling, clean-up, and sample collection.
  • Optimized for glycoprotein quantity up to 40 μg.
  • Rapid workflow: deglycosylation <15 min, labeling 5 min, clean-up 10 min.