Learn More
Waters Corp Waters N-Glycan Starter Kit, RapiFluor-MS, 24 Sample, HILIC
Catalog No. 501384498 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Starter Kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies sample preparation to three easy steps, completing the process in as little as 30 minutes, and includes components for deglycosylation, labeling, and clean-up.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Reduces sample preparation to 3 easy steps in 30 minutes.
  • RapiFluor-MS enhances fluorescence quantification and mass spectral response.
  • Single label for characterization and routine monitoring.
  • Processes 24 samples in a 3x8 format.
  • Optimized deglycosylation and efficient sample clean-up via SPE.
  • Reagent provides rapid tagging and enhanced ionization efficiency.
Catalog No. 50-138-4498 Supplier Waters Corp Supplier No. 176003712
The GlycoWorks RapiFluor-MS N-Glycan Starter Kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It simplifies sample preparation to three easy steps, completing the process in as little as 30 minutes, and includes components for deglycosylation, labeling, and clean-up.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Reduces sample preparation to 3 easy steps in 30 minutes.
  • RapiFluor-MS enhances fluorescence quantification and mass spectral response.
  • Single label for characterization and routine monitoring.
  • Processes 24 samples in a 3x8 format.
  • Optimized deglycosylation and efficient sample clean-up via SPE.
  • Reagent provides rapid tagging and enhanced ionization efficiency.