Learn More
Waters Corp Waters GlycoWorks RapiFluor-MS N-Glycan Basic Kit, 24 Sample, HILIC
Catalog No. 501384579 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Basic Kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. Sample preparation is simplified to three easy steps in as little as 30 minutes. The RapiFluor-MS reagent's unique properties provide increased fluorescence quantification and mass spectral response.
  • Streamlined sample preparation in 30 minutes.
  • Enhanced sensitivity with RapiFluor-MS for fluorescence quantification and mass spectral response.
  • Single label for comprehensive characterization and routine monitoring.
  • Improves product quality understanding via in-depth characterization.
  • Facilitates faster decision-making for N-linked glycans.
  • Includes deglycosylation, labeling, and clean-up modules.
Catalog No. 50-138-4579 Supplier Waters Corp Supplier No. 176003911
The GlycoWorks RapiFluor-MS N-Glycan Basic Kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. Sample preparation is simplified to three easy steps in as little as 30 minutes. The RapiFluor-MS reagent's unique properties provide increased fluorescence quantification and mass spectral response.
  • Streamlined sample preparation in 30 minutes.
  • Enhanced sensitivity with RapiFluor-MS for fluorescence quantification and mass spectral response.
  • Single label for comprehensive characterization and routine monitoring.
  • Improves product quality understanding via in-depth characterization.
  • Facilitates faster decision-making for N-linked glycans.
  • Includes deglycosylation, labeling, and clean-up modules.