Learn More
Waters Corp Waters GlycoWorks RapiFluor-MS N-Glycan Basic Kit, 96 Sample
Catalog No. 501384578 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This basic kit is designed to streamline and simplify N-glycan sample preparation. It allows for analysis in three easy steps, taking as little as 30 minutes. The RapiFluor-MS technology provides increased fluorescence quantification and enhanced mass spectral response, enabling the use of a single label for characterization to routine monitoring.
  • Simplifies and speeds up N-glycan sample preparation to just 3 easy steps in as little as 30 minutes.
  • Utilizes RapiFluor-MS technology for enhanced fluorescence quantification and mass spectral response.
  • Allows for sensitive glycan analysis and comprehensive characterization using a single label.
  • Processes 96 samples in a 4x24 format.
  • Reduces complicated, time-consuming sample preparation.
  • Enables easier and more sensitive glycan analysis.
  • Provides in-depth characterization of low abundance N-glycans.
Catalog No. 50-138-4578 Supplier Waters Corp Supplier No. 176003910
Add to Cart
Add to Cart
Request a Quote
This basic kit is designed to streamline and simplify N-glycan sample preparation. It allows for analysis in three easy steps, taking as little as 30 minutes. The RapiFluor-MS technology provides increased fluorescence quantification and enhanced mass spectral response, enabling the use of a single label for characterization to routine monitoring.
  • Simplifies and speeds up N-glycan sample preparation to just 3 easy steps in as little as 30 minutes.
  • Utilizes RapiFluor-MS technology for enhanced fluorescence quantification and mass spectral response.
  • Allows for sensitive glycan analysis and comprehensive characterization using a single label.
  • Processes 96 samples in a 4x24 format.
  • Reduces complicated, time-consuming sample preparation.
  • Enables easier and more sensitive glycan analysis.
  • Provides in-depth characterization of low abundance N-glycans.