Learn More
Waters Corp Waters Corporation Test Standard, RapiFluor-MS, High Mannose, N-Glycan Analysis
Catalog No. 501384793 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The RapiFluor-MS High Mannose Test Standard is for proficiency and system suitability testing in N-glycan analysis. It contains approximately 1000 pμmoles of purified, RapiFluor-MS labeled N-glycans from bovine ribonuclease B (M5 to M9 mannose structures). It aids in reliable detection and quantification of high mannose glycans for consistent glycan profiling.
  • Proficiency and system suitability standard for N-glycan analysis.
  • Contains approx. 1000 pμmoles of purified, RapiFluor-MS labeled N-glycans from bovine ribonuclease B.
  • Detects high mannose structures: M5 to M9.
  • Usable independently or spiked into N-glycan pools.
  • Ensures accuracy and consistency in glycan profiling.
  • Enhances signal intensity and improves glycan identification via RapiFluor-MS labeling.
  • Validates method performance, calibrates instruments, and verifies system suitability.
Catalog No. 50-138-4793 Supplier Waters Corp Supplier No. 186008317
The RapiFluor-MS High Mannose Test Standard is for proficiency and system suitability testing in N-glycan analysis. It contains approximately 1000 pμmoles of purified, RapiFluor-MS labeled N-glycans from bovine ribonuclease B (M5 to M9 mannose structures). It aids in reliable detection and quantification of high mannose glycans for consistent glycan profiling.
  • Proficiency and system suitability standard for N-glycan analysis.
  • Contains approx. 1000 pμmoles of purified, RapiFluor-MS labeled N-glycans from bovine ribonuclease B.
  • Detects high mannose structures: M5 to M9.
  • Usable independently or spiked into N-glycan pools.
  • Ensures accuracy and consistency in glycan profiling.
  • Enhances signal intensity and improves glycan identification via RapiFluor-MS labeling.
  • Validates method performance, calibrates instruments, and verifies system suitability.