Learn More
Waters Corp GlycoWorks HILIC Cartridge, 1 cc, 10 mg Sorbent, 20/pk
Catalog No. NC1768132 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks HILIC 1 cc Cartridge is used in glycoprotein sample preparation, facilitating the extraction of released glycans after denaturation and deglycosylation, and the removal of excess labeling reagent post-FLR labeling. It is specifically designed for protocols enriching FLR labeled glycans via reductive amination.
  • Extracts released glycans
  • Removes excess labeling reagent
  • Enriches FLR labeled glycans through reductive amination
  • Provides guidelines for N-linked glycan extraction, purification, and concentration, validated using IgG
  • Features a 1 cc cartridge with 10 mg sorbent
  • Packaged as 20 cartridges per package
  • Supports 10 reactions for 2-AB or 20 for RapiFluor-MS
  • Store at room temperature – dry
  • Minimum 1-year shelf life when unopened
Catalog No. NC1768132 Supplier Waters Corp Supplier No. 186007080
The GlycoWorks HILIC 1 cc Cartridge is used in glycoprotein sample preparation, facilitating the extraction of released glycans after denaturation and deglycosylation, and the removal of excess labeling reagent post-FLR labeling. It is specifically designed for protocols enriching FLR labeled glycans via reductive amination.
  • Extracts released glycans
  • Removes excess labeling reagent
  • Enriches FLR labeled glycans through reductive amination
  • Provides guidelines for N-linked glycan extraction, purification, and concentration, validated using IgG
  • Features a 1 cc cartridge with 10 mg sorbent
  • Packaged as 20 cartridges per package
  • Supports 10 reactions for 2-AB or 20 for RapiFluor-MS
  • Store at room temperature – dry
  • Minimum 1-year shelf life when unopened