Learn More
Waters Corp GlycoWorks Rapi-Fluor-MS N-Glycan Eco Basic Kit, 24 samples
Catalog No. 502886193 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GlycoWorks RapiFluor-MS N-Glycan Eco Kit streamlines N-glycan analysis with three easy, 30-minute steps. It uses RapiFluor-MS for enhanced fluorescence quantification and superior mass spectral response, and replaces DMF with a greener, safer DMSO solvent. The kit processes 24 samples.
  • Streamlined N-glycan analysis with three easy, 30-minute steps.
  • Enhanced fluorescence and mass spectral response with RapiFluor-MS.
  • Eco-friendly kit replaces DMF with DMSO and processes 24 samples.
Catalog No. 50-288-6193 Supplier Waters Corp Supplier No. 176005292
This GlycoWorks RapiFluor-MS N-Glycan Eco Kit streamlines N-glycan analysis with three easy, 30-minute steps. It uses RapiFluor-MS for enhanced fluorescence quantification and superior mass spectral response, and replaces DMF with a greener, safer DMSO solvent. The kit processes 24 samples.
  • Streamlined N-glycan analysis with three easy, 30-minute steps.
  • Enhanced fluorescence and mass spectral response with RapiFluor-MS.
  • Eco-friendly kit replaces DMF with DMSO and processes 24 samples.