Learn More
Waters Corp N-Glycan Eco Starter Kit, Premier UPLC Glycan BEH Column, 24 Samples
Catalog No. 502886257 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This starter kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It reduces complex sample preparation to three easy steps in as little as 30 minutes. The kit includes components for deglycosylation, labeling, clean-up, sample collection, a UPLC Glycan BEH Amide Column, and Mobile-Phase Concentrate for 24 samples.
  • Streamlined, fast, and highly sensitive analysis of released N-glycans.
  • Sample preparation reduced to 3 easy steps in as little as 30 minutes.
  • RapiFluor-MS provides increased fluorescence quantification and supreme mass spectral response.
  • Single label for characterization and routine monitoring.
  • Kit contains materials to process 24 samples in a 3x8 format.
Catalog No. 50-288-6257 Supplier Waters Corp Supplier No. 176005290
This starter kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans. It reduces complex sample preparation to three easy steps in as little as 30 minutes. The kit includes components for deglycosylation, labeling, clean-up, sample collection, a UPLC Glycan BEH Amide Column, and Mobile-Phase Concentrate for 24 samples.
  • Streamlined, fast, and highly sensitive analysis of released N-glycans.
  • Sample preparation reduced to 3 easy steps in as little as 30 minutes.
  • RapiFluor-MS provides increased fluorescence quantification and supreme mass spectral response.
  • Single label for characterization and routine monitoring.
  • Kit contains materials to process 24 samples in a 3x8 format.