Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Eco Refill Kit, 24 samples
Catalog No. 502886179 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans, simplifying sample preparation into three easy steps achievable in as little as 30 minutes. It utilizes RapiFluor-MS (RFMS) for enhanced fluorescence quantification and superior mass spectral response, enabling comprehensive characterization and routine monitoring with a single label. This Eco version replaces DMF with DMSO as a greener reagent solvent and contains all necessary components to process 24 samples.
  • Reduced sample preparation to 3 steps in as little as 30 minutes.
  • Enhanced sensitivity with RapiFluor-MS for superior fluorescence quantification and mass spectral response.
  • Eco-friendly reagent solvent, utilizing DMSO instead of DMF.
  • Processes 24 samples in a 3x8 format.
  • Ideal for eco-friendly, released N-linked glycan analysis.
Catalog No. 50-288-6179 Supplier Waters Corp Supplier No. 176005293
This kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans, simplifying sample preparation into three easy steps achievable in as little as 30 minutes. It utilizes RapiFluor-MS (RFMS) for enhanced fluorescence quantification and superior mass spectral response, enabling comprehensive characterization and routine monitoring with a single label. This Eco version replaces DMF with DMSO as a greener reagent solvent and contains all necessary components to process 24 samples.
  • Reduced sample preparation to 3 steps in as little as 30 minutes.
  • Enhanced sensitivity with RapiFluor-MS for superior fluorescence quantification and mass spectral response.
  • Eco-friendly reagent solvent, utilizing DMSO instead of DMF.
  • Processes 24 samples in a 3x8 format.
  • Ideal for eco-friendly, released N-linked glycan analysis.