Learn More
Waters Corp GlycoWorks Rapi-Fluor-MS N-Glycan Eco Kit, 24 Samples, N-Linked Glycan Analysis
Catalog No. 502886181 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Eco-friendly, released N-linked glycan analysis.
  • Processes 24 samples.
  • Reduces sample preparation time to as little as 30 min.
  • Uses RapiFluor-MS (RFMS) for increased fluorescence quantification and supreme mass spectral response.
  • Replaces DMF with DMSO as a greener reagent solvent.
  • Includes GlycoWorks Deglycosylation Module, GlycoWorks Eco Labeling Module, GlycoWorks Eco Clean-up Module, and GlycoWorks Sample Collection Module.
Catalog No. 50-288-6181 Supplier Waters Corp Supplier No. 176005291
  • Eco-friendly, released N-linked glycan analysis.
  • Processes 24 samples.
  • Reduces sample preparation time to as little as 30 min.
  • Uses RapiFluor-MS (RFMS) for increased fluorescence quantification and supreme mass spectral response.
  • Replaces DMF with DMSO as a greener reagent solvent.
  • Includes GlycoWorks Deglycosylation Module, GlycoWorks Eco Labeling Module, GlycoWorks Eco Clean-up Module, and GlycoWorks Sample Collection Module.