Learn More
Waters Corp Application Kit, Rapi-Fluor-MS, N-Glycan, Eco Starter Kit, 24 Samples
Catalog No. 502886210 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans in three easy steps, reducing sample preparation to as little as 30 minutes. It uses RapiFluor-MS for increased fluorescence quantification and superior mass spectral response. The Eco Kit utilizes a greener DMSO alternative as a reagent solvent and processes 24 samples.
  • Streamlined N-glycan analysis.
  • Three-step preparation in under 30 minutes.
  • Enhanced fluorescence and mass spectral response with RapiFluor-MS.
  • Eco-friendly with safer DMSO solvent.
  • Processes 24 samples.
  • Optimized deglycosylation and rapid labeling.
  • Efficient SPE clean-up.
  • Minimal sample handling.
  • Includes mass check standard.
  • Minimum 1 year shelf life.
Catalog No. 50-288-6210 Supplier Waters Corp Supplier No. 176005289
This kit offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans in three easy steps, reducing sample preparation to as little as 30 minutes. It uses RapiFluor-MS for increased fluorescence quantification and superior mass spectral response. The Eco Kit utilizes a greener DMSO alternative as a reagent solvent and processes 24 samples.
  • Streamlined N-glycan analysis.
  • Three-step preparation in under 30 minutes.
  • Enhanced fluorescence and mass spectral response with RapiFluor-MS.
  • Eco-friendly with safer DMSO solvent.
  • Processes 24 samples.
  • Optimized deglycosylation and rapid labeling.
  • Efficient SPE clean-up.
  • Minimal sample handling.
  • Includes mass check standard.
  • Minimum 1 year shelf life.