Learn More
Waters Corp Waters Buffer, Glycoworks Rapid, 5 mL
Catalog No. NC1732188 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Glycoworks Rapid Buffer, 5 mL, is a supplemental part number intended for customers utilizing the GlycoWorks RapiFluor-MS N-Glycan-96 and -24 sample kits. It is a concentrated, bulk solution of the customized rapid buffer used in the suggested deglycosylation method. This buffer is provided for customers who require buffer exchange before running their samples with the kit.
  • Streamlines purchasing with customized quotes.
  • Provides a concentrated, bulk solution of the customized rapid buffer used in the suggested deglycosylation method for N-Glycan-96 and -24 sample kits.
  • Designed for customers who need to perform buffer exchange before running their samples with the kit.
Catalog No. NC1732188 Supplier Waters Corp Supplier No. 186008100
The Glycoworks Rapid Buffer, 5 mL, is a supplemental part number intended for customers utilizing the GlycoWorks RapiFluor-MS N-Glycan-96 and -24 sample kits. It is a concentrated, bulk solution of the customized rapid buffer used in the suggested deglycosylation method. This buffer is provided for customers who require buffer exchange before running their samples with the kit.
  • Streamlines purchasing with customized quotes.
  • Provides a concentrated, bulk solution of the customized rapid buffer used in the suggested deglycosylation method for N-Glycan-96 and -24 sample kits.
  • Designed for customers who need to perform buffer exchange before running their samples with the kit.