Learn More
Waters Corp WATERS CORPORATION GlycoWorks Rapid Deglycosylation Kit, 1 x 24 Reactions
Catalog No. 501462018 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks Rapid Deglycosylation Kit is a convenient, versatile, and effective glycan sample preparation lab equipment. Designed to support RapiFluor-MS (RFMS) customers, it facilitates deglycosylation efficiency qualification and validation. The kit supports a 96-sample protocol, providing reagents for one 24-reaction sample set, and employs HILIC for separation.
  • Supports RapiFluor-MS (RFMS) deglycosylation efficiency qualification and validation.
  • Enables comparative analysis of intact and partial PNGase F deglycosylation.
  • Facilitates intact O-linked analysis with the ACQUITY UPLC Glycoprotein BEH Amide, 300A column.
  • Includes reagents for one 24-reaction sample set.
  • Key components: Rapid PNGaseF Enzyme, Glycoworks Rapid Buffer, and RapiGest SF Surfactant.
  • Employs HILIC for separation of isolated glycans.
  • Compatible with HRMS, MS, MS/MS, and UV detection methods.
Catalog No. 50-146-2018 Supplier Waters Corp Supplier No. 186008939
The GlycoWorks Rapid Deglycosylation Kit is a convenient, versatile, and effective glycan sample preparation lab equipment. Designed to support RapiFluor-MS (RFMS) customers, it facilitates deglycosylation efficiency qualification and validation. The kit supports a 96-sample protocol, providing reagents for one 24-reaction sample set, and employs HILIC for separation.
  • Supports RapiFluor-MS (RFMS) deglycosylation efficiency qualification and validation.
  • Enables comparative analysis of intact and partial PNGase F deglycosylation.
  • Facilitates intact O-linked analysis with the ACQUITY UPLC Glycoprotein BEH Amide, 300A column.
  • Includes reagents for one 24-reaction sample set.
  • Key components: Rapid PNGaseF Enzyme, Glycoworks Rapid Buffer, and RapiGest SF Surfactant.
  • Employs HILIC for separation of isolated glycans.
  • Compatible with HRMS, MS, MS/MS, and UV detection methods.