Learn More
Waters Corp Waters Corporation Deglycosylation Kit, 24 Samples, Sample Preparation Workflow, Glycan
Catalog No. 501461888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks Rapid Deglycosylation Kits enable validation of deglycosylation efficiency and comparison of intact with partial PNGase F deglycosylation for site occupancy and O-linked analysis. This kit supports a 24-sample protocol.
  • Validates deglycosylation efficiency.
  • Compares intact and partial PNGase F deglycosylation.
  • Facilitates intact O-linked analysis.
  • Provides reagents for 24 samples.
  • Streamlines N-glycan analysis by speeding deglycosylation and simplifying labeling.
  • Includes Rapid PNGaseF enzyme, Rapid Buffer, and RapiGest™ SF Surfactant.
  • Maximizes information generation during MS detection.
  • Offers MS-compatible HILIC based separations.
  • Used for glycoprotein heterogeneity removal and quality control.
Catalog No. 50-146-1888 Supplier Waters Corp Supplier No. 186008841
The GlycoWorks Rapid Deglycosylation Kits enable validation of deglycosylation efficiency and comparison of intact with partial PNGase F deglycosylation for site occupancy and O-linked analysis. This kit supports a 24-sample protocol.
  • Validates deglycosylation efficiency.
  • Compares intact and partial PNGase F deglycosylation.
  • Facilitates intact O-linked analysis.
  • Provides reagents for 24 samples.
  • Streamlines N-glycan analysis by speeding deglycosylation and simplifying labeling.
  • Includes Rapid PNGaseF enzyme, Rapid Buffer, and RapiGest™ SF Surfactant.
  • Maximizes information generation during MS detection.
  • Offers MS-compatible HILIC based separations.
  • Used for glycoprotein heterogeneity removal and quality control.