Learn More
Waters Corp GlycoWorks RapiFluo-MS N-Glycan Eco Starter Kit, 96 Samples, UPLC Glycan BEH Column
Catalog No. 502886200 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluo-MS N-Glycan Eco Starter Kit offers a fast, sensitive method for released N-glycan analysis. It simplifies sample preparation to three steps in 30 minutes. Using RapiFluor-MS (RFMS) technology, it provides increased fluorescence and superior mass spectral response for characterization and monitoring with a single label. The “Eco” design replaces DMF with DMSO as a safer reagent solvent.
  • Reduces N-glycan sample preparation to 3 easy steps in as little as 30 minutes.
  • Provides increased fluorescence quantification and superior mass spectral response with RapiFluor-MS technology.
  • Enables versatile labeling, using a single label for both characterization and routine monitoring.
  • Replaces DMF with DMSO as a greener and safer reagent solvent, making it eco-friendly.
  • Includes all necessary components for N-glycan analysis.
Catalog No. 50-288-6200 Supplier Waters Corp Supplier No. 176005286
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluo-MS N-Glycan Eco Starter Kit offers a fast, sensitive method for released N-glycan analysis. It simplifies sample preparation to three steps in 30 minutes. Using RapiFluor-MS (RFMS) technology, it provides increased fluorescence and superior mass spectral response for characterization and monitoring with a single label. The “Eco” design replaces DMF with DMSO as a safer reagent solvent.
  • Reduces N-glycan sample preparation to 3 easy steps in as little as 30 minutes.
  • Provides increased fluorescence quantification and superior mass spectral response with RapiFluor-MS technology.
  • Enables versatile labeling, using a single label for both characterization and routine monitoring.
  • Replaces DMF with DMSO as a greener and safer reagent solvent, making it eco-friendly.
  • Includes all necessary components for N-glycan analysis.