Learn More
Waters Corp WATERS CORPORATION GlycoWorks RapiFluor-MS N-Glycan Eco Starter Kit, 96 Samples
Catalog No. 502886232 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed to provide scientists with all the necessary materials to successfully perform fast N-glycan deglycosylation, rapid fluorescent labeling, and glycan extraction with minimal preparation time. It uses optimized deglycosylation conditions and reagents, a novel rapid label called RapiFluor-MS for sensitive fluorescence and mass detection, and solid-phase extraction (SPE) for sample clean-up prior to LC analyses.
  • Optimized deglycosylation conditions and reagents for fast release.
  • Features RapiFluor-MS, a novel rapid label for sensitive fluorescence and mass detection.
  • Sample clean-up via solid-phase extraction (SPE) with the HILIC μElution™ Plate.
  • Streamlined protocol for fast sample preparation, minimizing handling and loss.
  • Each reaction is optimized for the release, labeling, and extraction of N-glycans from 15 to 40 μg of glycoprotein.
Catalog No. 50-288-6232 Supplier Waters Corp Supplier No. 176005285
Add to Cart
Add to Cart
Request a Quote
This kit is designed to provide scientists with all the necessary materials to successfully perform fast N-glycan deglycosylation, rapid fluorescent labeling, and glycan extraction with minimal preparation time. It uses optimized deglycosylation conditions and reagents, a novel rapid label called RapiFluor-MS for sensitive fluorescence and mass detection, and solid-phase extraction (SPE) for sample clean-up prior to LC analyses.
  • Optimized deglycosylation conditions and reagents for fast release.
  • Features RapiFluor-MS, a novel rapid label for sensitive fluorescence and mass detection.
  • Sample clean-up via solid-phase extraction (SPE) with the HILIC μElution™ Plate.
  • Streamlined protocol for fast sample preparation, minimizing handling and loss.
  • Each reaction is optimized for the release, labeling, and extraction of N-glycans from 15 to 40 μg of glycoprotein.