Learn More
Waters Corp Waters Labeling Module, RapiFluor-MS, N-Glycan Analysis, 96-Sample
Catalog No. 501528582 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS Labeling Module is a 96-sample kit for N-glycan analysis. It uses RapiFluor-MS to enhance fluorescence quantification and mass spectral response, streamlining sample preparation for analysis in as little as 30 minutes.
  • Enhanced fluorescence quantification and mass spectral response
  • Single label for comprehensive N-glycan analysis and monitoring
  • Streamlined preparation for rapid deglycosylation, labeling, and clean-up in <30 minutes
  • Dual detection capabilities for versatile analysis
  • Optimized reagents ensure efficient N-glycan release and high yield
  • Robust clean-up via HILIC μElution Plate
  • High throughput, designed for 96 samples
  • Mass spectrometry compatibility for direct LC-MS analysis
Catalog No. 50-152-8582 Supplier Waters Corp Supplier No. 186007989
The GlycoWorks RapiFluor-MS Labeling Module is a 96-sample kit for N-glycan analysis. It uses RapiFluor-MS to enhance fluorescence quantification and mass spectral response, streamlining sample preparation for analysis in as little as 30 minutes.
  • Enhanced fluorescence quantification and mass spectral response
  • Single label for comprehensive N-glycan analysis and monitoring
  • Streamlined preparation for rapid deglycosylation, labeling, and clean-up in <30 minutes
  • Dual detection capabilities for versatile analysis
  • Optimized reagents ensure efficient N-glycan release and high yield
  • Robust clean-up via HILIC μElution Plate
  • High throughput, designed for 96 samples
  • Mass spectrometry compatibility for direct LC-MS analysis