Learn More
Waters Corp Waters Corporation Labeling Module, RapiFluor-MS, Automation, GlycoWorks, 48 or 96 Samples
Catalog No. 502038839 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS Labeling Module-Automation provides all the components needed for RapiFluor-MS labeling for automation platforms. It is designed for quick enzymatic release and labeling of N-glycans and can process 48 or 96 samples at a time, allowing for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal human participation.
  • Provides all components for RapiFluor-MS labeling on automation platforms, enabling quick enzymatic release and labeling of N-glycans.
  • Processes 48 or 96 samples for high-throughput glycoprotein preparation.
  • Features a fast label for sensitive fluorescence and strong mass detection.
  • Includes two 55 mg vials of RapiFluor-MS reagent and two 1 mL vials of reagent solvent.
  • Contains hazardous material, requiring special freight handling.
Catalog No. 50-203-8839 Supplier Waters Corp Supplier No. 186008822
The GlycoWorks RapiFluor-MS Labeling Module-Automation provides all the components needed for RapiFluor-MS labeling for automation platforms. It is designed for quick enzymatic release and labeling of N-glycans and can process 48 or 96 samples at a time, allowing for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal human participation.
  • Provides all components for RapiFluor-MS labeling on automation platforms, enabling quick enzymatic release and labeling of N-glycans.
  • Processes 48 or 96 samples for high-throughput glycoprotein preparation.
  • Features a fast label for sensitive fluorescence and strong mass detection.
  • Includes two 55 mg vials of RapiFluor-MS reagent and two 1 mL vials of reagent solvent.
  • Contains hazardous material, requiring special freight handling.