Learn More
Waters Corp Waters Labeling Module, RapiFluor-MS, N-Glycan Analysis, 24-Sample
Catalog No. 501461886 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This compact and effective labeling module, part of the GlycoWorks RapiFluor-MS N-Glycan Kit, contains all reagents for rapid, streamlined, and highly sensitive N-glycan analysis. It simplifies sample preparation, enabling analysis in 30 minutes with three easy steps. The labeling reagent offers unique chemical attributes for increased fluorescence and supreme mass spectral response, enhancing glycan analysis and product quality understanding.
  • Rapid N-glycan analysis.
  • Streamlined and highly sensitive.
  • Reduces sample preparation time.
  • Enables analysis in 30 minutes with 3 steps.
  • Increased fluorescence quantification.
  • Supreme mass spectral response.
  • Single label for characterization to routine monitoring.
  • Improves product quality understanding.
  • Facilitates faster identification of low abundance N-linked glycans.
Catalog No. 50-146-1886 Supplier Waters Corp Supplier No. 186008091
This compact and effective labeling module, part of the GlycoWorks RapiFluor-MS N-Glycan Kit, contains all reagents for rapid, streamlined, and highly sensitive N-glycan analysis. It simplifies sample preparation, enabling analysis in 30 minutes with three easy steps. The labeling reagent offers unique chemical attributes for increased fluorescence and supreme mass spectral response, enhancing glycan analysis and product quality understanding.
  • Rapid N-glycan analysis.
  • Streamlined and highly sensitive.
  • Reduces sample preparation time.
  • Enables analysis in 30 minutes with 3 steps.
  • Increased fluorescence quantification.
  • Supreme mass spectral response.
  • Single label for characterization to routine monitoring.
  • Improves product quality understanding.
  • Facilitates faster identification of low abundance N-linked glycans.