Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit, Automation, 96 Analyses, HILIC
Catalog No. 502390833 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit – Automation enables high-throughput glycoprotein sample preparation for N-glycan analysis on automated platforms. Its protocol is validated for monoclonal antibodies and diverse N-linked glycoproteins, with scaled reagents for versatile automated instrumentation.
  • Includes script parameters guideline and liquid classes table to accelerate automated script development.
  • Contains RFMS labeled standard, column, and mobile phase concentrate for RapiFluor-MS Library compatibility.
  • Supports 96 analyses, featuring HILIC separation and MS, HRMS, MS/MS, UV detection methods.
  • Components requiring cold chain shipping (deglycosylation module, standards) are clearly designated.
  • Optimized for LC-MS systems in glycan applications.
Catalog No. 50-239-0833 Supplier Waters Corp Supplier No. 176005282
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit – Automation enables high-throughput glycoprotein sample preparation for N-glycan analysis on automated platforms. Its protocol is validated for monoclonal antibodies and diverse N-linked glycoproteins, with scaled reagents for versatile automated instrumentation.
  • Includes script parameters guideline and liquid classes table to accelerate automated script development.
  • Contains RFMS labeled standard, column, and mobile phase concentrate for RapiFluor-MS Library compatibility.
  • Supports 96 analyses, featuring HILIC separation and MS, HRMS, MS/MS, UV detection methods.
  • Components requiring cold chain shipping (deglycosylation module, standards) are clearly designated.
  • Optimized for LC-MS systems in glycan applications.