Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit, Automation, 96 Analyses
Catalog No. 502390805 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit – Automation enables high-throughput glycoprotein sample preparation for N-glycan analysis on automated platforms. Validated for monoclonal antibodies and various other N-linked glycoproteins, it streamlines workflows for glycan detection with unmatched fluorescent and mass spectrometric performance.
  • Designed for automated, high-throughput sample prep
  • Validated with mAbs and compatible with N-linked glycoproteins
  • Reagents scaled for automated instruments, offering versatility
  • Includes script guidelines and liquid class table
  • Label provides high mass spectral signal for low abundance glycans
  • Contains deglycosylation, labeling, clean-up, and sample collection modules
  • Supports 96 analyses
Catalog No. 50-239-0805 Supplier Waters Corp Supplier No. 176004150
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Script Parameters Starter Kit – Automation enables high-throughput glycoprotein sample preparation for N-glycan analysis on automated platforms. Validated for monoclonal antibodies and various other N-linked glycoproteins, it streamlines workflows for glycan detection with unmatched fluorescent and mass spectrometric performance.
  • Designed for automated, high-throughput sample prep
  • Validated with mAbs and compatible with N-linked glycoproteins
  • Reagents scaled for automated instruments, offering versatility
  • Includes script guidelines and liquid class table
  • Label provides high mass spectral signal for low abundance glycans
  • Contains deglycosylation, labeling, clean-up, and sample collection modules
  • Supports 96 analyses