Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Kit, Automation, 96 Analysis, Sample Preparation Workflow
Catalog No. 502391869 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Kit – Automation is designed for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. This protocol has been validated using monoclonal antibodies and has also been tested for a wide range of other N-linked glycoproteins. All reagents in the kit are scaled to residual volumes needed for many automated instrumentation, allowing for versatility when choosing your platform. The kit is in a 2 x 48 format.
  • Designed for automated platforms, enabling high-throughput glycoprotein sample preparation.
  • Minimizes user intervention.
  • Protocol validated with monoclonal antibodies and tested for a wide range of other N-linked glycoproteins.
  • Reagents scaled to residual volumes for versatility across automated instrumentation.
  • Kit format: 2 x 48.
Catalog No. 50-239-1869 Supplier Waters Corp Supplier No. 176004153
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Kit – Automation is designed for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. This protocol has been validated using monoclonal antibodies and has also been tested for a wide range of other N-linked glycoproteins. All reagents in the kit are scaled to residual volumes needed for many automated instrumentation, allowing for versatility when choosing your platform. The kit is in a 2 x 48 format.
  • Designed for automated platforms, enabling high-throughput glycoprotein sample preparation.
  • Minimizes user intervention.
  • Protocol validated with monoclonal antibodies and tested for a wide range of other N-linked glycoproteins.
  • Reagents scaled to residual volumes for versatility across automated instrumentation.
  • Kit format: 2 x 48.