Learn More
Waters Corp N-Glycan Script Starter Kit, Automated, 96 Analysis, HILIC
Catalog No. 502392062 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Script Starter Kit-Automation is designed for high-throughput glycoprotein sample preparation and N-glycan analysis using automated platforms. This kit enables minimal user intervention and is validated with monoclonal antibodies and various N-linked glycoproteins.
  • Designed for automated platforms
  • Enables high-throughput glycoprotein sample preparation
  • Supports N-glycan analysis
  • Requires minimal user intervention
  • Protocol validated using monoclonal antibodies
  • Tested for a wide range of other N-linked glycoproteins
  • Reagents scaled for automated instrumentation versatility
  • Kit provided in a 2 x 48 format
  • Includes CD and Workstation requirements document with script
  • Includes RFMS labeled standard, column, and mobile phase concentrate for RapiFluor-MS Library use
Catalog No. 50-239-2062 Supplier Waters Corp Supplier No. 176004151
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Script Starter Kit-Automation is designed for high-throughput glycoprotein sample preparation and N-glycan analysis using automated platforms. This kit enables minimal user intervention and is validated with monoclonal antibodies and various N-linked glycoproteins.
  • Designed for automated platforms
  • Enables high-throughput glycoprotein sample preparation
  • Supports N-glycan analysis
  • Requires minimal user intervention
  • Protocol validated using monoclonal antibodies
  • Tested for a wide range of other N-linked glycoproteins
  • Reagents scaled for automated instrumentation versatility
  • Kit provided in a 2 x 48 format
  • Includes CD and Workstation requirements document with script
  • Includes RFMS labeled standard, column, and mobile phase concentrate for RapiFluor-MS Library use