Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Script Starter Kit, Automated, High-Throughput, 96 Analyses, LC-MS
Catalog No. 502392455 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for automated platforms, enabling high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. Validated with monoclonal antibodies, it’s effective for a wide range of N-linked glycoproteins. Reagents are scaled to residual volumes, providing versatility for automated instrumentation.
  • Automated platform compatibility.
  • High-throughput N-glycan analysis.
  • Minimal user intervention.
  • Validated for monoclonal antibodies and N-linked glycoproteins.
  • Reagents scaled for automated instrumentation.
  • Includes scripts for faster automation results.
  • 96 analyses (2 × 48 format).
  • HILIC separation mode.
  • Supports MS, HRMS, MS/MS, and UV detection.
  • LC-MS system type.
Catalog No. 50-239-2455 Supplier Waters Corp Supplier No. 176005283
Add to Cart
Add to Cart
Request a Quote
This kit is designed for automated platforms, enabling high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. Validated with monoclonal antibodies, it’s effective for a wide range of N-linked glycoproteins. Reagents are scaled to residual volumes, providing versatility for automated instrumentation.
  • Automated platform compatibility.
  • High-throughput N-glycan analysis.
  • Minimal user intervention.
  • Validated for monoclonal antibodies and N-linked glycoproteins.
  • Reagents scaled for automated instrumentation.
  • Includes scripts for faster automation results.
  • 96 analyses (2 × 48 format).
  • HILIC separation mode.
  • Supports MS, HRMS, MS/MS, and UV detection.
  • LC-MS system type.