Learn More
Waters Corp N-Glycan Eco Starter Kit, Automation, RapiFluor-MS, 2 x 48 Format
Catalog No. 502886196 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Eco Starter Kit – Automation is designed for automated platforms, enabling high-throughput glycoprotein sample preparation for N-glycan analysis. It is validated for monoclonal antibodies and other N-linked glycoproteins. This kit uses DMSO as a greener, safer alternative for the RapiFluor-MS Reagent Label, replacing DMF.
  • Designed for automated glycoprotein sample preparation.
  • Validated for monoclonal antibodies and N-linked glycoproteins.
  • Reagents scaled for automated instrumentation.
  • Replaces DMF with greener DMSO for RapiFluor-MS Reagent Label.
  • Removes other DMF-containing solutions.
  • Includes RFMS labeled standard, column, and mobile phase concentrate.
  • Kit format: 2 x 48.
  • Suggested use: Eco-friendly, released N-linked glycan analysis, automation.
Catalog No. 50-288-6196 Supplier Waters Corp Supplier No. 176005302
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Eco Starter Kit – Automation is designed for automated platforms, enabling high-throughput glycoprotein sample preparation for N-glycan analysis. It is validated for monoclonal antibodies and other N-linked glycoproteins. This kit uses DMSO as a greener, safer alternative for the RapiFluor-MS Reagent Label, replacing DMF.
  • Designed for automated glycoprotein sample preparation.
  • Validated for monoclonal antibodies and N-linked glycoproteins.
  • Reagents scaled for automated instrumentation.
  • Replaces DMF with greener DMSO for RapiFluor-MS Reagent Label.
  • Removes other DMF-containing solutions.
  • Includes RFMS labeled standard, column, and mobile phase concentrate.
  • Kit format: 2 x 48.
  • Suggested use: Eco-friendly, released N-linked glycan analysis, automation.