Learn More
Waters Corp N-Glycan Starter Kit, RapiFluor-MS, 96 Samples
Catalog No. 502886199 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This starter kit provides a streamlined and highly sensitive method for N-glycan analysis. It simplifies sample preparation into three easy steps, completed in as little as 30 minutes. The RapiFluor-MS technology offers increased fluorescence and superior mass spectral response, enabling comprehensive glycan characterization and routine monitoring.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Sample preparation in three easy steps, completed in as little as 30 minutes.
  • RapiFluor-MS technology for increased fluorescence and superior mass spectral response.
  • Processes 96 samples in a 4x24 format.
  • Includes deglycosylation, labeling, clean-up, and sample collection modules.
  • Features a UPLC glycan BEH amide column and mobile-phase concentrate.
Catalog No. 50-288-6199 Supplier Waters Corp Supplier No. 176005243
Add to Cart
Add to Cart
Request a Quote
This starter kit provides a streamlined and highly sensitive method for N-glycan analysis. It simplifies sample preparation into three easy steps, completed in as little as 30 minutes. The RapiFluor-MS technology offers increased fluorescence and superior mass spectral response, enabling comprehensive glycan characterization and routine monitoring.
  • Streamlined, fast, and highly sensitive N-glycan analysis.
  • Sample preparation in three easy steps, completed in as little as 30 minutes.
  • RapiFluor-MS technology for increased fluorescence and superior mass spectral response.
  • Processes 96 samples in a 4x24 format.
  • Includes deglycosylation, labeling, clean-up, and sample collection modules.
  • Features a UPLC glycan BEH amide column and mobile-phase concentrate.