Learn More
Waters Corp N-Glycan Kit, Eco Basic, 96 Samples
Catalog No. 502886256 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GlycoWorks RapiFluor-MS N-Glycan Eco Kit simplifies and accelerates glycan analysis sample preparation, reducing it to 30 minutes in three easy steps. Utilizing RapiFluor-MS (RFMS) for enhanced fluorescence quantification and superior mass spectral response, it allows a single label for characterization and routine monitoring. The “Eco” version uses greener DMSO as a reagent solvent, replacing DMF.
  • Reduces sample preparation to 30 minutes in 3 easy steps.
  • Enhanced detection provides increased fluorescence quantification and supreme mass spectral response.
  • Single labeling technology can be used for both glycan characterization and routine monitoring.
  • Eco-friendly as it replaces DMF with DMSO as a greener and safer solvent.
  • Includes deglycosylation, eco labeling, and eco clean-up modules.
Catalog No. 50-288-6256 Supplier Waters Corp Supplier No. 176005288
Add to Cart
Add to Cart
Request a Quote
This GlycoWorks RapiFluor-MS N-Glycan Eco Kit simplifies and accelerates glycan analysis sample preparation, reducing it to 30 minutes in three easy steps. Utilizing RapiFluor-MS (RFMS) for enhanced fluorescence quantification and superior mass spectral response, it allows a single label for characterization and routine monitoring. The “Eco” version uses greener DMSO as a reagent solvent, replacing DMF.
  • Reduces sample preparation to 30 minutes in 3 easy steps.
  • Enhanced detection provides increased fluorescence quantification and supreme mass spectral response.
  • Single labeling technology can be used for both glycan characterization and routine monitoring.
  • Eco-friendly as it replaces DMF with DMSO as a greener and safer solvent.
  • Includes deglycosylation, eco labeling, and eco clean-up modules.