Learn More
Waters Corp GlycoWorks RapiFluor-MS N-glycan Eco Labeling Kit, 24 Samples
Catalog No. 502886178 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for eco-friendly released N-linked glycan analysis and is intended to be used as part of a larger GlycoWorks RapiFluor-MS N-Glycan Eco Kit. It utilizes a unique chemical label that provides increased fluorescence quantification and superior mass spectral response, allowing for both characterization and routine monitoring with a single label. The kit replaces DMF with DMSO as a greener, safer alternative for the reagent solvent.
  • Increased fluorescence quantification
  • Superior mass spectral response
  • Suitable for both characterization and routine monitoring
  • Eco-friendly solvent replacement (DMSO instead of DMF)
  • Compatible with UPLC/UHPLC/HPLC separation modes
  • Supports HRMS, MS, MS/MS, and UV detection methods
  • Each vial of labeling reagent is suitable for 24 reactions
Catalog No. 50-288-6178 Supplier Waters Corp Supplier No. 186010608
This kit is designed for eco-friendly released N-linked glycan analysis and is intended to be used as part of a larger GlycoWorks RapiFluor-MS N-Glycan Eco Kit. It utilizes a unique chemical label that provides increased fluorescence quantification and superior mass spectral response, allowing for both characterization and routine monitoring with a single label. The kit replaces DMF with DMSO as a greener, safer alternative for the reagent solvent.
  • Increased fluorescence quantification
  • Superior mass spectral response
  • Suitable for both characterization and routine monitoring
  • Eco-friendly solvent replacement (DMSO instead of DMF)
  • Compatible with UPLC/UHPLC/HPLC separation modes
  • Supports HRMS, MS, MS/MS, and UV detection methods
  • Each vial of labeling reagent is suitable for 24 reactions