Learn More
Waters Corp GlycoWorks RapiFluor-MS N-glycan Eco Labeling Kit, Automation
Catalog No. 502886226 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor–MS Labeling Module–Automation provides all the components needed for RapiFluor–MS labeling used for automation platforms. GlycoWorks RapiFluor–MS N–Glycan Eco Kits replace DMF with a greener, safer, alternative (DMSO) as a reagent solvent for the RapiFluor–MS reagent label.
  • Replaces DMF with greener, safer alternative (DMSO) as reagent solvent for RapiFluor–MS reagent label
  • Designed for automation platforms
  • Eco–friendly for released N–linked glycan analysis
  • Sample preparation workflow
  • Compatible with UPLC/UHPLC/HPLC separation modes
  • Supports HRMS, MS, MS/MS, and UV detection methods
  • Suitable for LC–MS system type
  • Room temperature storage and shipping
Catalog No. 50-288-6226 Supplier Waters Corp Supplier No. 186010609
The GlycoWorks RapiFluor–MS Labeling Module–Automation provides all the components needed for RapiFluor–MS labeling used for automation platforms. GlycoWorks RapiFluor–MS N–Glycan Eco Kits replace DMF with a greener, safer, alternative (DMSO) as a reagent solvent for the RapiFluor–MS reagent label.
  • Replaces DMF with greener, safer alternative (DMSO) as reagent solvent for RapiFluor–MS reagent label
  • Designed for automation platforms
  • Eco–friendly for released N–linked glycan analysis
  • Sample preparation workflow
  • Compatible with UPLC/UHPLC/HPLC separation modes
  • Supports HRMS, MS, MS/MS, and UV detection methods
  • Suitable for LC–MS system type
  • Room temperature storage and shipping