Learn More
Waters Corp Waters GlycoWorks Kit, RapiFluor-MS N-Glycan, Automation, ACQUITY Glycan Column
Catalog No. 502886229 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters GlycoWorks RapiFluor-MS N-Glycan Eco Script Parameters Starter Kit with ACQUITY Premier Glycan BEH Amide Column — Automation is designed for fast enzymatic release and rapid labeling of N-glycans. It facilitates high-throughput glycoprotein sample preparation on automated platforms, processing 48 or 96 samples. The kit includes optimized deglycosylation conditions, RapiFluor-MS for sensitive fluorescence and enhanced mass detection, and a GlycoWorks HILIC μElution Plate for sample cleanup. Full automation for 96 samples can be completed in under 3 hours.
  • Fast enzymatic release and rapid labeling of N-glycans.
  • Scaled to process 48 or 96 samples for high-throughput automation.
  • Contains RapiFluor-MS for sensitive fluorescence and enhanced mass detection.
  • Includes solid-phase extraction (SPE) for sample cleanup.
Catalog No. 50-288-6229 Supplier Waters Corp Supplier No. 176005299
Add to Cart
Add to Cart
Request a Quote
The Waters GlycoWorks RapiFluor-MS N-Glycan Eco Script Parameters Starter Kit with ACQUITY Premier Glycan BEH Amide Column — Automation is designed for fast enzymatic release and rapid labeling of N-glycans. It facilitates high-throughput glycoprotein sample preparation on automated platforms, processing 48 or 96 samples. The kit includes optimized deglycosylation conditions, RapiFluor-MS for sensitive fluorescence and enhanced mass detection, and a GlycoWorks HILIC μElution Plate for sample cleanup. Full automation for 96 samples can be completed in under 3 hours.
  • Fast enzymatic release and rapid labeling of N-glycans.
  • Scaled to process 48 or 96 samples for high-throughput automation.
  • Contains RapiFluor-MS for sensitive fluorescence and enhanced mass detection.
  • Includes solid-phase extraction (SPE) for sample cleanup.