Learn More
Waters Corp N-Glycan Eco Script Starter Kit, GlycoWorks RapiFluor-MS, Automation, 2 x 48 Format
Catalog No. 502886230 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Eco Script Starter Kit – Automation is engineered for high-throughput glycoprotein sample preparation on automated platforms, requiring minimal user involvement for N-glycan analysis.
  • Includes a CD and workstation requirements document containing the script developed during verification testing for automated systems.
  • Contains an RFMS labeled standard, column, and mobile phase concentrate recommended for use with the RapiFluor-MS library.
Catalog No. 50-288-6230 Supplier Waters Corp Supplier No. 176005300
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Eco Script Starter Kit – Automation is engineered for high-throughput glycoprotein sample preparation on automated platforms, requiring minimal user involvement for N-glycan analysis.
  • Includes a CD and workstation requirements document containing the script developed during verification testing for automated systems.
  • Contains an RFMS labeled standard, column, and mobile phase concentrate recommended for use with the RapiFluor-MS library.