Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Starter Kit, 24 Samples, ACQUITY Premier UPLC Glycan BEH Column
Catalog No. 502886212 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides everything necessary for enzymatic deglycosylation, RapiFluor-MS labeling, SPE clean-up, and UPLC analysis, including a UPLC column and mobile phase concentrate. It offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans.
  • Three easy steps in as little as 30 minutes, reducing complicated sample preparation.
  • RapiFluor-MS provides increased fluorescence quantification and supreme mass spectral response.
  • Uses a single label for characterization to routine monitoring.
  • Processes 24 samples in a 3×8 format.
  • Includes a labeling module for the solubilization and labeling of released glycosylamine.
  • Features a UPLC Glycan BEH Amide column, 1.7 μm, 2.1 × 150 mm.
Catalog No. 50-288-6212 Supplier Waters Corp Supplier No. 176005242
This kit provides everything necessary for enzymatic deglycosylation, RapiFluor-MS labeling, SPE clean-up, and UPLC analysis, including a UPLC column and mobile phase concentrate. It offers a streamlined, fast, and highly sensitive method for analyzing released N-glycans.
  • Three easy steps in as little as 30 minutes, reducing complicated sample preparation.
  • RapiFluor-MS provides increased fluorescence quantification and supreme mass spectral response.
  • Uses a single label for characterization to routine monitoring.
  • Processes 24 samples in a 3×8 format.
  • Includes a labeling module for the solubilization and labeling of released glycosylamine.
  • Features a UPLC Glycan BEH Amide column, 1.7 μm, 2.1 × 150 mm.