Learn More
Waters Corp GlycoWorks N-Glycan Eco Kit, RapiFluor-MS, 96 Samples
Catalog No. 502886213 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-glycan Eco Kit provides a streamlined, fast, and highly sensitive method for analyzing released N-glycans, simplifying sample preparation into three steps in as little as 30 minutes. It utilizes RapiFluor-MS for enhanced fluorescence quantification and supreme mass spectral response, enabling a single label for both characterization and routine monitoring. This Eco Kit replaces DMF with anhydrous DMSO as a greener and safer reagent solvent.
  • Streamlined, fast, sensitive N-glycan analysis.
  • Three-step sample prep in as little as 30 minutes.
  • RapiFluor-MS for enhanced fluorescence and mass spectral response.
  • Single label for characterization and routine monitoring.
  • Eco-friendly; uses anhydrous DMSO instead of DMF.
  • Processes 96 samples in a 4x24 format.
  • Supports UPLC/UHPLC/HPLC separation; HRMS, MS, MS/MS, UV detection.
Catalog No. 50-288-6213 Supplier Waters Corp Supplier No. 176005287
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-glycan Eco Kit provides a streamlined, fast, and highly sensitive method for analyzing released N-glycans, simplifying sample preparation into three steps in as little as 30 minutes. It utilizes RapiFluor-MS for enhanced fluorescence quantification and supreme mass spectral response, enabling a single label for both characterization and routine monitoring. This Eco Kit replaces DMF with anhydrous DMSO as a greener and safer reagent solvent.
  • Streamlined, fast, sensitive N-glycan analysis.
  • Three-step sample prep in as little as 30 minutes.
  • RapiFluor-MS for enhanced fluorescence and mass spectral response.
  • Single label for characterization and routine monitoring.
  • Eco-friendly; uses anhydrous DMSO instead of DMF.
  • Processes 96 samples in a 4x24 format.
  • Supports UPLC/UHPLC/HPLC separation; HRMS, MS, MS/MS, UV detection.