Learn More
Waters Corp GlycoWorks N-Glycan Kit, RapiFluor-MS, Automation, Eco Basic
Catalog No. 502886231 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Kit – Automation provides materials for fast enzymatic release and rapid labeling of N-glycans, designed for automated, high-throughput glycoprotein sample preparation and N-glycan analysis.
  • Automated processing for 48 or 96 samples.
  • Rapid enzymatic release and dual-detection labeling with RapiFluor-MS.
  • Efficient Solid-Phase Extraction (SPE) cleanup via HILIC μElution™ Plate.
  • Expedited workflow: full automation of 96 samples in less than 3 hours.
  • Includes RapiFluor-MS reagent, RapiGest SF surfactant, and Rapid PNGase F enzyme.
  • Compatible with direct LC-MS analysis; optimized for 15 to 40 μg glycoproteins.
  • Validated for monoclonal antibodies and various N-linked glycoproteins.
Catalog No. 50-288-6231 Supplier Waters Corp Supplier No. 176005305
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Kit – Automation provides materials for fast enzymatic release and rapid labeling of N-glycans, designed for automated, high-throughput glycoprotein sample preparation and N-glycan analysis.
  • Automated processing for 48 or 96 samples.
  • Rapid enzymatic release and dual-detection labeling with RapiFluor-MS.
  • Efficient Solid-Phase Extraction (SPE) cleanup via HILIC μElution™ Plate.
  • Expedited workflow: full automation of 96 samples in less than 3 hours.
  • Includes RapiFluor-MS reagent, RapiGest SF surfactant, and Rapid PNGase F enzyme.
  • Compatible with direct LC-MS analysis; optimized for 15 to 40 μg glycoproteins.
  • Validated for monoclonal antibodies and various N-linked glycoproteins.