Learn More
Waters Corp GlycoWorks RapiFluor-MS N-glycan Eco Labeling Kit, 96 samples, Eco-Friendly
Catalog No. 502886240 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-glycan Eco Labeling Kit is designed for 96 samples, utilizing RapiFluor-MS technology for increased fluorescence quantification and superior mass spectral response. This kit allows for a single label for glycan characterization and routine monitoring, replacing DMF with DMSO as a greener, safer reagent solvent.
  • Designed for 96 analyses.
  • Utilizes a sample preparation workflow.
  • Supports UPLC/UHPLC/HPLC separation modes.
  • Compatible with HRMS, MS, MS/MS, and UV detection methods.
  • Storage and shipping at room temperature.
  • Ideal for glycan applications.
  • Suggested for eco-friendly, released N-linked glycan analysis.
  • Contents include 4 labeling reagents and 4 reagent solvents (DMSO).
  • Provides increased fluorescence quantification and supreme mass spectral response.
Catalog No. 50-288-6240 Supplier Waters Corp Supplier No. 186010607
The GlycoWorks RapiFluor-MS N-glycan Eco Labeling Kit is designed for 96 samples, utilizing RapiFluor-MS technology for increased fluorescence quantification and superior mass spectral response. This kit allows for a single label for glycan characterization and routine monitoring, replacing DMF with DMSO as a greener, safer reagent solvent.
  • Designed for 96 analyses.
  • Utilizes a sample preparation workflow.
  • Supports UPLC/UHPLC/HPLC separation modes.
  • Compatible with HRMS, MS, MS/MS, and UV detection methods.
  • Storage and shipping at room temperature.
  • Ideal for glycan applications.
  • Suggested for eco-friendly, released N-linked glycan analysis.
  • Contents include 4 labeling reagents and 4 reagent solvents (DMSO).
  • Provides increased fluorescence quantification and supreme mass spectral response.