Learn More
Waters Corp WATERS CORPORATION RapiFluor-MS N-Glycan Kit, Automation, 96 Samples
Catalog No. 502886245 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS N-Glycan Kits for automation are designed for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. The protocol is validated for monoclonal antibodies and a wide range of other N-linked glycoproteins.
  • Rapid sample preparation in three steps, under an hour.
  • Enhanced fluorescence quantification and superior mass spectral response with RapiFluor-MS labeling.
  • Single label for characterization and routine monitoring.
  • Comprehensive kits for deglycosylation, labeling, and SPE cleanup.
  • Reagents scaled for automated instruments, offering platform versatility.
  • Includes script information to accelerate automation results.
  • “Eco” version uses DMSO as the reagent solvent.
  • HILIC μElution Plate for efficient contaminant removal.
Catalog No. 50-288-6245 Supplier Waters Corp Supplier No. 176005298
Add to Cart
Add to Cart
Request a Quote
The GlycoWorks RapiFluor-MS N-Glycan Kits for automation are designed for high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. The protocol is validated for monoclonal antibodies and a wide range of other N-linked glycoproteins.
  • Rapid sample preparation in three steps, under an hour.
  • Enhanced fluorescence quantification and superior mass spectral response with RapiFluor-MS labeling.
  • Single label for characterization and routine monitoring.
  • Comprehensive kits for deglycosylation, labeling, and SPE cleanup.
  • Reagents scaled for automated instruments, offering platform versatility.
  • Includes script information to accelerate automation results.
  • “Eco” version uses DMSO as the reagent solvent.
  • HILIC μElution Plate for efficient contaminant removal.