Learn More
Waters Corp GlycoWorks RapiFluor-MS N-Glycan Eco Script Starter Kit, ACQUITY Premier Glycan BEH Column, Automation
Catalog No. 502886246 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit enables automated, high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. Validated with monoclonal antibodies, it supports various N-linked glycoproteins. Reagents are scaled for automated platforms, offering versatility. Eco kits replace DMF with greener DMSO solvent.
  • Automated, high-throughput glycoprotein sample preparation.
  • Fast enzymatic release and rapid N-glycan labeling.
  • Novel rapid label for sensitive fluorescence and mass detection.
  • Efficient solid-phase extraction cleanup.
  • Processes 96 samples in under 3 hours when automated.
  • Eco-friendly formulation with DMSO solvent.
  • Includes deglycosylation, labeling, cleanup, and collection modules.
  • Supplied with glycan BEH Amide Column and mobile phase concentrate.
Catalog No. 50-288-6246 Supplier Waters Corp Supplier No. 176005301
Add to Cart
Add to Cart
Request a Quote
This kit enables automated, high-throughput glycoprotein sample preparation for N-glycan analysis with minimal user intervention. Validated with monoclonal antibodies, it supports various N-linked glycoproteins. Reagents are scaled for automated platforms, offering versatility. Eco kits replace DMF with greener DMSO solvent.
  • Automated, high-throughput glycoprotein sample preparation.
  • Fast enzymatic release and rapid N-glycan labeling.
  • Novel rapid label for sensitive fluorescence and mass detection.
  • Efficient solid-phase extraction cleanup.
  • Processes 96 samples in under 3 hours when automated.
  • Eco-friendly formulation with DMSO solvent.
  • Includes deglycosylation, labeling, cleanup, and collection modules.
  • Supplied with glycan BEH Amide Column and mobile phase concentrate.