Learn More
Waters Corp N-Glycan Sample Preparation Kit, Eco-Friendly, Automation, 2 x 48 Format
Catalog No. 502886251 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit enables high-throughput glycoprotein sample preparation for N-glycan analysis, ideal for automated platforms with minimal user intervention. Validated with monoclonal antibodies and effective for various N-linked glycoproteins, this eco-friendly kit replaces DMF with DMSO and removes other DMF-containing solutions. It is provided in a 2 x 48 format.
  • Rapid labeling for sensitive fluorescence detection and strong mass signal intensity.
  • Streamlined protocol with fast deglycosylation, labeling, and cleanup steps.
  • Designed for full automation on compatible platforms, processing 96 samples in under 3 hours.
  • Optimized reagents ensure efficient deglycosylation and prevent precipitation.
  • Efficient cleanup through solid-phase extraction removes reaction by-products.
Catalog No. 50-288-6251 Supplier Waters Corp Supplier No. 176005304
Add to Cart
Add to Cart
Request a Quote
This kit enables high-throughput glycoprotein sample preparation for N-glycan analysis, ideal for automated platforms with minimal user intervention. Validated with monoclonal antibodies and effective for various N-linked glycoproteins, this eco-friendly kit replaces DMF with DMSO and removes other DMF-containing solutions. It is provided in a 2 x 48 format.
  • Rapid labeling for sensitive fluorescence detection and strong mass signal intensity.
  • Streamlined protocol with fast deglycosylation, labeling, and cleanup steps.
  • Designed for full automation on compatible platforms, processing 96 samples in under 3 hours.
  • Optimized reagents ensure efficient deglycosylation and prevent precipitation.
  • Efficient cleanup through solid-phase extraction removes reaction by-products.