Learn More
Waters Corp Waters Corporation Kit, GlycoWorks RapiFluor-MS, N-Glycan, Basic, Automation
Catalog No. 502037564 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for automated, high-throughput glycoprotein sample preparation, enabling N-glycan analysis with minimal user intervention. It is validated for monoclonal antibodies and tested for various N-linked glycoproteins. Reagents are scaled for compatibility with automated instrumentation, offering versatility in platform choice. This basic version excludes the Sample Collection Module, allowing users to integrate their own consumables for automation platforms.
  • 96 analysis.
  • Sample preparation workflow.
  • HILIC separation mode.
  • MS, MS/MS, UV, HRMS detection method.
  • Glycan application.
  • 1 unit per package.
Catalog No. 50-203-7564 Supplier Waters Corp Supplier No. 176004154
Add to Cart
Add to Cart
Request a Quote
This kit is designed for automated, high-throughput glycoprotein sample preparation, enabling N-glycan analysis with minimal user intervention. It is validated for monoclonal antibodies and tested for various N-linked glycoproteins. Reagents are scaled for compatibility with automated instrumentation, offering versatility in platform choice. This basic version excludes the Sample Collection Module, allowing users to integrate their own consumables for automation platforms.
  • 96 analysis.
  • Sample preparation workflow.
  • HILIC separation mode.
  • MS, MS/MS, UV, HRMS detection method.
  • Glycan application.
  • 1 unit per package.