Learn More
Waters Corp GlycoWorks Sample Preparation Kit, Reductive Amination, High-throughput, 96-Well Plate
Catalog No. 502391161 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks Glycan Sample Preparation Kits are designed to generate high-quality data from analyses of pre-column derivatized, labeled glycans, synergistically with the ACQUITY UPLC Glycan Application Solution. This kit employs HILIC Solid Phase Extraction (SPE) material to separate released glycans from protein and labeled glycans from excess free label. It includes a 96-well plate for high-throughput applications.
  • Includes a 96-well plate and is configured for high-throughput applications.
  • Utilizes HILIC Solid Phase Extraction (SPE) material for glycan separation.
  • Contains a 1 mg vial of RapiGest SF to aid in protein deglycosylation.
  • Includes a control standard vial to confirm correct protocol performance.
  • Comes with a reagent kit.
  • Designed for 96 analyses.
  • Uses MS, MS/MS, UV, and HRMS detection methods.
  • Suitable for LC-MS system type.
Catalog No. 50-239-1161 Supplier Waters Corp Supplier No. 176003090
The GlycoWorks Glycan Sample Preparation Kits are designed to generate high-quality data from analyses of pre-column derivatized, labeled glycans, synergistically with the ACQUITY UPLC Glycan Application Solution. This kit employs HILIC Solid Phase Extraction (SPE) material to separate released glycans from protein and labeled glycans from excess free label. It includes a 96-well plate for high-throughput applications.
  • Includes a 96-well plate and is configured for high-throughput applications.
  • Utilizes HILIC Solid Phase Extraction (SPE) material for glycan separation.
  • Contains a 1 mg vial of RapiGest SF to aid in protein deglycosylation.
  • Includes a control standard vial to confirm correct protocol performance.
  • Comes with a reagent kit.
  • Designed for 96 analyses.
  • Uses MS, MS/MS, UV, and HRMS detection methods.
  • Suitable for LC-MS system type.